Skip to Content
MilliporeSigma
Allopurinol Related Compound E Pharmaceutical Secondary Standard; Certified Reference Material

Allopurinol Related Compound E

Allopurinol Related Compound E Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Ethyl 3-(formylamino)-1H-pyrazole-4-carboxylate, Ethyl 5-(formylamino)-1H-pyrazole-4-carboxylate
Empirical Formula (Hill Notation):
C7H9N3O3
CAS Number:
Molecular Weight:
183.16

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade