Skip to Content
MilliporeSigma
(4S,4′S)-4,4′-[[4-(Dimethylamino)butylidene]bis[[3-[2-(dimethylamino)ethyl]-1H-indole-2,5-diyl]methylene]]bis[2-oxazolidinone] pharmaceutical secondary standard, certified reference material

(4S,4′S)-4,4′-[[4-(Dimethylamino)butylidene]bis[[3-[2-(dimethylamino)ethyl]-1H-indole-2,5-diyl]methylene]]bis[2-oxazolidinone]

(4S,4′S)-4,4′-[[4-(Dimethylamino)butylidene]bis[[3-[2-(dimethylamino)ethyl]-1H-indole-2,5-diyl]methylene]]bis[2-oxazolidinone] pharmaceutical secondary standard, certified reference material
Synonym(s):
2,2′-[4-(Dimethylamino)butane-1,1-diyl]bis{5-[(S)-(2-oxooxazolidin-4-yl)methyl]-3-(2-dimethylaminoethyl)indole}, Zolmitriptan dimer
Empirical Formula (Hill Notation):
C38H53N7O4
CAS Number:
Molecular Weight:
671.87

Products

Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade