Skip to Content
MilliporeSigma
4-((6-Amino-5-bromo-2-((4-cyanophenyl)amino)pyrimidin-4-yl)oxy)-3,5-dimethylbenzonitrile

4-((6-Amino-5-bromo-2-((4-cyanophenyl)amino)pyrimidin-4-yl)oxy)-3,5-dimethylbenzonitrile

4-((6-Amino-5-bromo-2-((4-cyanophenyl)amino)pyrimidin-4-yl)oxy)-3,5-dimethylbenzonitrile
Empirical Formula (Hill Notation):
C20H15BrN6O
CAS Number:
Molecular Weight:
435.28

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Showing 1-1 of 1