Skip to Content
MilliporeSigma
4,4′-Diaminodiphenyl sulfide analytical standard

4,4′-Diaminodiphenyl sulfide

4,4′-Diaminodiphenyl sulfide analytical standard
Synonym(s):
4,4′-Thiodianiline
Linear Formula:
S(C6H4NH2)2
CAS Number:
Molecular Weight:
216.30
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing