Skip to Content
MilliporeSigma
2-Tridecanone 99%

2-Tridecanone

2-Tridecanone 99%
Synonym(s):
Methyl undecyl ketone
Linear Formula:
CH3(CH2)10COCH3
CAS Number:
Molecular Weight:
198.34
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing