![2-Deoxy-α-D-ribose 1-phosphate bis(cyclohexylammonium) salt ≥97% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/127/533/be8bd9ae-3a8c-488a-93b4-98c6482ce0ac/640/be8bd9ae-3a8c-488a-93b4-98c6482ce0ac.png)
2-Deoxy-α-D-ribose 1-phosphate bis(cyclohexylammonium) salt
![2-Deoxy-α-D-ribose 1-phosphate bis(cyclohexylammonium) salt ≥97% (TLC)](/deepweb/assets/sigmaaldrich/product/structures/127/533/be8bd9ae-3a8c-488a-93b4-98c6482ce0ac/640/be8bd9ae-3a8c-488a-93b4-98c6482ce0ac.png)
Empirical Formula (Hill Notation):
C5H11O7P · 2C6H13N
CAS Number:
Molecular Weight:
412.46
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing