Skip to Content
MilliporeSigma
1,2,3,4-Tetrahydro-4,6-dihydroxy-2-methylisoquinoline hydrochloride Pharmaceutical Secondary Standard; Certified Reference Material

1,2,3,4-Tetrahydro-4,6-dihydroxy-2-methylisoquinoline hydrochloride

1,2,3,4-Tetrahydro-4,6-dihydroxy-2-methylisoquinoline hydrochloride Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
2-Methyl-1,2,3,4-tetrahydro-4,6-isoquinolinediol hydrochloride
Empirical Formula (Hill Notation):
C10H13NO2 · HCl
CAS Number:
Molecular Weight:
215.68

Products

Brand
Formula Weight
Melting Point (°C)
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade