Skip to Content
MilliporeSigma
All Photos(1)

Documents

23230-U

Supelco

Amber storage bottles

volume 120 mL, O.D. × H 49 mm × 114 mm, thread for 22-400, PTFE/silicone septum, black phenolic hole cap, pkg of 12 ea

Synonym(s):

SCREW-CAP AMBER SEPTUM BOTTLE 120ML PK12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.11

material

PTFE septum face
PTFE/silicone septum
amber glass bottle
black phenolic hole cap

feature

closure type screw top bottle
pre-assembled

packaging

pkg of 12 ea

O.D. × H

49 mm × 114 mm

volume

120 mL

fitting

thread for 22-400

Looking for similar products? Visit Product Comparison Guide

General description

Amber storage bottles are primarily used to protect the stored sample from sunlight. Oxidation of various oils is prevented when packaged in amber glass bottles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service