Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1377

Sigma-Aldrich

R-7050

≥95% (HPLC)

Synonym(s):

8-Chloro-4-(phenylsulfanyl)-1-(trifluoromethyl)[1,2,4]triazolo[4,3-a]quinoxaline, 8-Chloro-4-(phenylthio)-1-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]quinoxaline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H8ClF3N4S
CAS Number:
Molecular Weight:
380.77
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

shipped in

wet ice

storage temp.

−20°C

SMILES string

ClC1=CC=C(N=C(SC2=CC=CC=C2)C3=NN=C(C(F)(F)F)N34)C4=C1

InChI

1S/C16H8ClF3N4S/c17-9-6-7-11-12(8-9)24-13(22-23-15(24)16(18,19)20)14(21-11)25-10-4-2-1-3-5-10/h1-8H

InChI key

SUUMKHOVGVYGOP-UHFFFAOYSA-N

Biochem/physiol Actions

R-7050 is a cell-permeable TNF-α receptor antagonist that blocks TNF-α-induced binding of TNF-αRI with TNFαR-associated death domain protein and receptor interacting protein 1, blocking internalization of the TNFα-TNFαR complex. R-7050 was found to reduce neurovascular injury in a mouse model of hemorrhagic stroke.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Mudersbach et al.
PloS one, 14(5), e0216218-e0216218 (2019-05-03)
The angiotensin-converting enzyme (ACE) plays a central role in the renin-angiotensin system, which is involved in the regulation of blood pressure. Alterations in ACE expression or activity are associated with various pathological phenotypes, particularly cardiovascular diseases. In human endothelial cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service