Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00270

Sigma-Aldrich

Trachelosperogenin A1

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H56O12
CAS Number:
Molecular Weight:
680.82
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

C[C@@H]1CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)C[C@@H](O)[C@H](O)[C@@](C)([C@@H]5CC[C@@]34C)C(O)=O)[C@@H]2[C@]1(C)O)C(=O)O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O

InChI

1S/C36H56O12/c1-17-9-12-36(30(45)48-28-25(41)24(40)23(39)20(16-37)47-28)14-13-32(3)18(26(36)35(17,6)46)7-8-21-31(2)15-19(38)27(42)34(5,29(43)44)22(31)10-11-33(21,32)4/h7,17,19-28,37-42,46H,8-16H2,1-6H3,(H,43,44)/t17-,19-,20-,21-,22-,23-,24+,25-,26-,27+,28+,31-,32-,33-,34-,35-,36+/m1/s1

InChI key

YXSQSVWHKZZWDD-IWSVBZIKSA-N

General description

Natural product derived from plant source.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service