Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1403651

Sigma-Aldrich

Monoclonal Anti-CD97 antibody produced in mouse

clone 3B11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

TM7LN1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3B11, monoclonal

form

buffered aqueous solution

mol wt

antigen ~107.73 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD97(976)

General description

This gene is a member of the EGF-TM7 family of class II seven-span transmembrane (7-TM) molecules, likely encoded by a gene cluster on the short arm of chromosome 19. The encoded product is a glycoprotein that is present on the surface of most activated leukocytes and spans the membrane seven times, which is a defining feature of G protein-coupled receptors. The protein has an extended extracellular region with several N-terminal epidermal growth factor (EGF)-like domains, which mediate binding to its cellular ligand, decay accelerating factor (DAF, CD55), a regulatory protein of the complement cascade. The presence of structural features characteristic of extracellular matrix proteins and transmembrane proteins suggests that this protein is a receptor involved in both cell adhesion and signaling processes early after leukocyte activation. Alternative splicing has been observed for this gene and three variants have been found. (provided by RefSeq)

Immunogen

CD97 (AAH26690, 1 a.a. ~ 742 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MGGRVFLAFCVWLTLPGAETQDSRGCARWCPQNSSCVNATACRCNPGFSSFSEIITTPTETCDDINECATPSKVSCGKFSDCWNTEGSYDCVCSPGYEPVSGAKTFKNESENTCQDVDECSSGRHQCDSSTVCFNTVGSYSCRCRPGWKPRHGIPNNQKDTVCEDMTFSTWTPPPGVHSQTLSRFFDKVQDLGRDSKTSSAEVTIQNVIKLVDELMEAPGDVEALAPPVRHLIATQLLSNLEDIMRILAKSLPKGPFTYISPSNTELTLMIQERGDKNVTMGQSSARMKLNWAVAAGAEDPGPAVAGILSIQNMTTLLANASLNLHSKKQAELEEIYESSIRGVQLRRLSAVNSIFLSHNNTKELNSPILFAFSHLESSDGEAGRDPPAKDVMPGPRQELLCAFWKSDSDRGGHWATEGCQVLGSKNGSTTCQCSHLSSFAILMAHYDVEDWKLTLITRVGLALSLFCLLLCILTFLLVRPIQGSRTTIHLHLCICLFVGSTIFLAGIENEGGQVGLRCRLVAGLLHYCFLAAFCWMSLEGLELYFLVVRVFQGQGLSTRWLCLIGYGVPLLIVGVSAAIYSKGYGRPRYCWLDFEQGFLWSFLGPVTFIILCNAVIFVTTVWKLTQKFSEINPDMKKLKKARALTITAIAQLFLLGCTWVFGLFIFDDRSLVLTYVFTILNCLQGAFLYLLHCLLNKKVREEYRKWACLVAGGSKYSEFTSTTSGTGHNQTRALRASESGI

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service