Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0261

Sigma-Aldrich

PF-06649283

≥98% (HPLC)

Synonym(s):

(4R,4aR,6R,8aS)-8a-(2,4-Difluoro-phenyl)-4-methyl-6-(3-methyl-isoxazol-5-yl)-4,4a,5,6,8,8a-hexahydro-7-oxa-3-thia-1-aza-naphthalen-2-ylamine, (4R,4aR,6R,8aS)-8a-(2,4-Difluorophenyl)-4,4a,5,6,8,8a-hexahydro-4-methyl-6-(3-methyl-5-isoxazolyl)-pyrano[3,4-d][1,3]thiazin-2-amine, PF06649283

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19F2N3O2S
CAS Number:
Molecular Weight:
379.42
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

C[C@@H]1[C@@]2([H])[C@@](CO[C@@H](C3=CC(C)=NO3)C2)(C4=CC=C(F)C=C4F)N=C(N)S1

InChI

1S/C18H19F2N3O2S/c1-9-5-16(25-23-9)15-7-13-10(2)26-17(21)22-18(13,8-24-15)12-4-3-11(19)6-14(12)20/h3-6,10,13,15H,7-8H2,1-2H3,(H2,21,22)/t10-,13+,15-,18-/m1/s1

InChI key

HVEVDFRWOQHAPJ-CKMNQQOVSA-N

Biochem/physiol Actions

PF-06649283 is a new β-Secretase (BACE) inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service