Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L5414

Sigma-Aldrich

Laser Tough-Spots®

diam. 3/8 in., blue

Synonym(s):

solvent resistant labels, temperature resistant labels

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
55121611
NACRES:
NB.35

material

blue polyolefin

packaging

pack of 3840 ea (192 labels per sheet)

manufacturer/tradename

Diversified Biotech SPOT-1100

diam.

3/8 in.

color

blue

Looking for similar products? Visit Product Comparison Guide

General description

Tough-Spots are chemically inert, polyvinyl labels that adhere to all plastics. Tough-Spots withstand boiling water baths, conventional and cryogenic freezer temperatures, most organic solvents, caustic agents and other challenges without peeling, becoming brittle or degrading. Labels can be printed directly from the laser printer. 20 sheets per pack.

Legal Information

Tough-Spots is a registered trademark of Diversified Biotech

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service