Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0000565

β-Acetyldigoxin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Digoxin 4″′-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H66O15
CAS Number:
Molecular Weight:
822.98
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

β-acetyldigoxin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O1CC(=CC1=O)C2C3(C(C4C(C5(C(CC4)CC(CC5)OC6OC(C(C(C6)O)OC7OC(C(C(C7)O)OC8OC(C(C(C8)O)OC(=O)C)C)C)C)C)CC3O)(CC2)O)C

InChI

1S/C43H66O15/c1-20-38(55-23(4)44)30(45)17-36(53-20)57-40-22(3)54-37(18-32(40)47)58-39-21(2)52-35(16-31(39)46)56-26-9-11-41(5)25(14-26)7-8-28-29(41)15-33(48)42(6)27(10-12-43(28,42)50)24-13-34(49)51-19-24/h13,20-22,25-33,35-40,45-48,50H,7-12,14-19H2,1-6H3

InChI key

NREAGDHHMSOWKZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

β-Acetyldigoxin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmaceutical Manufacturing Encyclopedia, 64-64 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service