Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR2790

Supelco

Fingolimod Hydrochloride

Synonym(s):

FTY720, 2-Amino-2-[2-(4-octyl-phenyl)-ethyl]-propane-1,3-diol hydrochloride, Fingolimod hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H33NO2 · HCl
CAS Number:
Molecular Weight:
343.93
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0002055
traceable to USP 1270480

description

Pharmaceutical Secondary Standard; Certified Reference Material

API family

fingolimod

form

powder

packaging

pkg of 500 mg

application(s)

pharmaceutical small molecule

storage temp.

2-30°C

SMILES string

Cl.NC(CO)(CO)CCc1ccc(cc1)CCCCCCCC

InChI

1S/C19H33NO2.ClH/c1-2-3-4-5-6-7-8-17-9-11-18(12-10-17)13-14-19(20,15-21)16-22;/h9-12,21-22H,2-8,13-16,20H2,1H3;1H

InChI key

SWZTYAVBMYWFGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

FTY720 is an immunomodulating drug and sphingosine 1-phosphate (S1P) receptor modulator. Phosphorylation of FTY270 by sphingosine kinase causes S1P1R internalization, which sequesters lymphocytes in lymph nodes, preventing them from taking part in an autoimmune response. Clinically, it has been approved for the treatment of multiple sclerosis (MS). It has also been shown to block and reverse paclitaxel-induced chemotherapy induced peripheral neuropathy (CIPN) through S1PR inhibition as well as inhibit the activity of histone deacetylases in the hippocampus of mouse brains, thereby modulating memory.
FTY720, an immunomodulating drug, acts as a sphingosine 1-phosphate (S1P) receptor modulator.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC6340 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2

Target Organs

Immune system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service