Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50374

Supelco

Methyl palmitate

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

n-Hexadecanoic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14CO2CH3
CAS Number:
Molecular Weight:
270.45
Beilstein:
1780973
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.25

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.4512 (lit.)

bp

185 °C/10 mmHg (lit.)

mp

32-35 °C (lit.)

density

0.852 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C17H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(18)19-2/h3-16H2,1-2H3

InChI key

FLIACVVOZYBSBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service