Skip to Content
MilliporeSigma
All Photos(1)

Documents

SMEQ00KT

Milli-Q

System Mounting Kit for POD of Milli-Q® EQ Systems

Designed for use with Milli-Q® EQ 7 series systems to mount the Q-POD® dispenser directly on the system.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
JA.13

feature

Allows to install and mount the Q-POD® dispenser on the system

manufacturer/tradename

Milli-Q®

system H

9.4 cm (3.7 in.)

system W

19.0 cm (7.4 in.)

system depth

18.6 cm (7.3 in.)

compatibility

for use with Milli-Q® EQ 7000
for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016

shipped in

ambient

General description

The system mounting kit for Milli-Q® EQ 7000/08/16 systems lets you achieve a more compact configuration by allowing you to place the Q-POD® dispenser directly on the system itself. You can position the POD in four places: on the top right or left, or on the bottom right or left. This way it is in the most convenient place adapted to your lab’s needs.

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Q-POD is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service