Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABE265

Sigma-Aldrich

Anti-phospho HuR (Ser221) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

ELAV-like protein 1, Hu-antigen R, HuR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), bovine (based on 100% sequence homology), chicken (based on 100% sequence homology), Xenopus (based on 100% sequence homology), mouse (based on 100% sequence homology)

technique(s)

inhibition assay: suitable (peptide)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer221)

Gene Information

human ... ELAVL1(1994)

General description

HuR , also known as Hu-antigen R, ELAV-like protein 1, and encoded by the gene name ELAVL1 and HUR, is a member of the RRM ELAV family. HuR is an RNA bindnig protein that contains 3 RNA-binding domains. It is known to binds AU-rich elements if mRNA. It regulates gene expression by destabilizing their mRNA. Recent studies have shown that HuR is upregulated in certain cancers such as breast and gastric cancers. HuR is also involved in embryonic stem cells (ESCs) differentiation. HuR preferentially binds mRNAs that are not methylated by N6-methyladenosine (m6A), stabilizing them, and thus promoting ESCs differentiation. Additionally, HuR has been shown to bind to poly-U elements and AU-rich elements (AREs) in the 3′-UTR of target mRNAs, as well as the AU-rich element in FOS and IL3/interleukin-3 mRNAs. Some of HuR’s targets include cFos, IL-3, and BRCA1 mRNAs. HuR is also involved in the regulation of VEFG-A and NFkB expression in certain cell types. HuR activation and accumulation are also linked to many energy signaling molcules such as that of the PI3K/Akt pathway and AMPK.

Immunogen

Epitope: Phosphorylated Ser221
KLH-conjugated linear peptide corresponding to human phospho HuR (Ser221).

Application

Anti-phospho HuR (Ser221) | ABE265 is an antibody against phospho HuR for use in Western Blotting, Peptide Inhibition Assay.
Peptide Inhibition Analysis: A 1:200 dilution from a representative lot blocked phospho HuR (Ser221) in HCT116 cell lysate.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors

Quality

Evaluated by Western Blotting in HCT116 cell lysate.

Western Blotting Analysis: A 1:200 dilution of this antibody detected phospho HuR (Ser221) in 10 µg of HCT116 cell lysate.

Target description

~37 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katherine Rothamel et al.
Cell reports, 35(8), 109178-109178 (2021-05-27)
Upon pathogen detection, the innate immune system triggers signaling events leading to upregulation of pro-inflammatory and anti-microbial mRNA transcripts. RNA-binding proteins (RBPs) interact with these critical mRNAs and regulate their fates at the post-transcriptional level. One such RBP is ELAVL1.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service