Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

568500

Sigma-Aldrich

SQ 22536

Cell-permeable adenylate cyclase inhibitor.

Synonym(s):

SQ 22536, 9-(Tetrahydro-2ʹ-furyl)adenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11N5O
CAS Number:
Molecular Weight:
205.22
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 25 mg/mL
water: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C9H11N5O/c10-8-7-9(12-4-11-8)14(5-13-7)6-2-1-3-15-6/h4-6H,1-3H2,(H2,10,11,12)

InChI key

UKHMZCMKHPHFOT-UHFFFAOYSA-N

General description

Cell-permeable adenylate cyclase inhibitor. Blocks PTH-stimulation of adenylate cyclase (IC50 = 200 µM). Reduces PGE2-induced inhibition of O2· production.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
adenylate cyclase
Product does not compete with ATP.
Reversible: no
Target IC50: 200 µM against adenylate cyclase

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 2 weeks at 4°C.

Other Notes

Talpain, E., et al. 1995. Br. J. Pharmacol. 114, 1459.
Tamaoki, J., et al. 1993. Prostaglandins45, 363.
Fabbri, E., et al. 1991. J. Enzyme Inhib.5, 87.
Goldsmith, B.A. and Abrams, T.W. 1991. Proc. Natl. Acad. Sci. USA 88, 9021.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magdalena Szymanska et al.
Scientific reports, 13(1), 14133-14133 (2023-08-30)
Angiogenesis is important for endometrial remodeling in mature females. The endometrium synthesizes high amounts of prostacyclin (PGI2) but the role of PGI2 in angiogenesis-related events in this tissue was not fully described. In the present study, porcine endometrial endothelial (pEETH)
NaHye Lee et al.
The FEBS journal, 287(17), 3841-3870 (2020-02-01)
Microglia (MG), the principal neuroimmune sentinels in the brain, continuously sense changes in their environment and respond to invading pathogens, toxins, and cellular debris, thereby affecting neuroinflammation. Microbial pathogens produce small metabolites that influence neuroinflammation, but the molecular mechanisms that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service