Skip to Content
MilliporeSigma

5.00849

SAFC

Vitamin A

water-miscible 1 ml = 200,000 I.U. for oral use, EMPROVE® ESSENTIAL, Ph. Eur., BP

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352401

Agency

BP
Ph. Eur.

Quality Level

product line

EMPROVE® ESSENTIAL

form

viscous liquid

pH

6.5-7.5 (20 °C, 100 g/L in H2O)

solubility

100 g/L

density

1.04 g/cm3 at 20 °C

application(s)

liquid formulation
pharmaceutical

storage temp.

15-25°C

General description

We offer a range of high quality critical raw materials, solvents and reagents plus the documentation and expertise to meet stringent regulatory requirements. In addition to our comprehensive portfolio, we also offer custom manufacturing services to meet your exact needs. As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Vitamin A is used in clinical nutrition for treatment of vitamin A deficiency and as skin care agent. Vitamin A Emprove® Essential is suitable for pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service