Key Documents
Z287261
Sigma-Aldrich
Aldrich® low-pressure three station manifold with fixed outlet pressure regulator
outlet NPTM 1/4 in.
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
outlet NPTM
1/4 in.
Related Categories
General description
This system offers complete manifold installation flexibility. As increased manifold capacity is needed, simply order the required parts and install them on-site. Made of brass with 1/4 in. NPTM inlet. Similar to basic system, but with a fixed-pressure regulator that limits gas pressure at the manifold outlets to less than 5 psig. This safety feature guards against accidental over-pressurization of fragile glass vessels, bottles, and reaction equipment.
Caution
Do not exceed 200 psi inlet gas pressure.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC