Key Documents
Z203750
Sigma-Aldrich
Aldrich® large-capacity cold trap
Inside finger for traps with ST/NS: 55/50 joint
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
Related Categories
General description
With removable cold-fingers and a choice of 55/50 joint or 50 mm O-ring joint. Traps with O-ring joint include pinch clamp. Use with or without dry ice to condense and collect in trap. Dewar flask F9651 is recommended for use with 500 mL capacity cold traps.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC