Documents
Z190543
Sigma-Aldrich
Liquid scintillation vials (with screw cap attached)
urethane foam-filled high-density polyethylene cap, liner (Poly-Seal cone), pkg of 500 ea
Synonym(s):
Scintillation Counting Vials
About This Item
UNSPSC Code:
41121806
NACRES:
NB.11
material
clear glass vial
liner (Poly-Seal cone)
urethane foam-filled high-density polyethylene cap
feature
closure type screw top vial
closure type white polyethylene cap
packaging
pkg of 500 ea
size
28 mm × 61 mm
volume
20 mL
fitting
thread for 22-400
General description
Autoclavable, easy to maintain. A vial is used to store or as a closure type,in order to prevent contamination.
Reliable Wheaton "180" brand low-potassium glass. White 22 mm caps have tops suitable for writing. Vials with caps attached are packed in individual cells in trays of 100.