Key Documents
Z126586
Sigma-Aldrich
Recessed septa mixed set
set of 1
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
material
rubber septa (silicone)
packaging
set of 1
General description
Contains ten of each size for the following septa, with carrying box:
Fitting Neck I.D.
8.7 mm
10.7 mm
15.0 mm
17.0 mm
23.0 mm