Key Documents
TEM-LC325CU
Sigma-Aldrich
Lacey Carbon Supported Copper Grids
size 300 mesh, size 50 nm , hole size, box of 25 ×
Synonym(s):
Lacey/Carbon Film, 300 Mesh, Copper, 5pk
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 25 ×
size
300 mesh
50 nm , hole size
General description
Lacey carbon film supported TEM grids. This carbon coating consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.