跳转至内容
Merck
  • Mature adipocytes in bone marrow protect myeloma cells against chemotherapy through autophagy activation.
Mature adipocytes in bone marrow protect myeloma cells against chemotherapy through autophagy activation.
Oncotarget (2015-10-13)
Zhiqiang Liu, Jingda Xu, Jin He, Huan Liu, Pei Lin, Xinhai Wan, Nora M Navone, Qiang Tong, Larry W Kwak, Robert Z Orlowski, Jing Yang
摘要
A major problem in patients with multiple myeloma is chemotherapy resistance, which develops in myeloma cells upon interaction with bone marrow stromal cells. However, few studies have determined the role of bone marrow adipocytes, a major component of stromal cells in the bone marrow, in myeloma chemotherapy resistance. We reveal that mature human adipocytes activate autophagy and upregulate the expression of autophagic proteins, thereby suppressing chemotherapy-induced caspase cleavage and apoptosis in myeloma cells. We found that adipocytes secreted known and novel adipokines, such as leptin and adipsin. The addition of these adipokines enhanced the expression of autophagic proteins and reduced apoptosis in myeloma cells. In vivo studies further demonstrated the importance of bone marrow-derived adipocytes in the reduced response of myeloma cells to chemotherapy. Our findings suggest that adipocytes, adipocyte-secreted adipokines, and adipocyte-activated autophagy are novel targets for combatting chemotherapy resistance and enhancing treatment efficacy in myeloma patients.
材料
货号
品牌
产品描述
Sigma-Aldrich
MISSION® esiRNA, targeting human ATG5
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Atg5