跳转至内容
Merck
  • Genomic organization and chromosomal localization of three members of the UDP-N-acetylgalactosamine: polypeptide N-acetylgalactosaminyltransferase family.
Genomic organization and chromosomal localization of three members of the UDP-N-acetylgalactosamine: polypeptide N-acetylgalactosaminyltransferase family.
Glycobiology (1998-06-11)
E P Bennett, D O Weghuis, G Merkx, A G van Kessel, H Eiberg, H Clausen
摘要
A homologous family of UDP- N -acetylgalactosamine: polypeptide N -acetylgalactosaminyltransferases (GalNAc-transferases) initiate O-glycosylation. These transferases share overall amino acid sequence similarities of approximately 45-50%, but segments with higher similarities of approximately 80% are found in the putative catalytic domain. Here we have characterized the genomic organization of the coding regions of three GalNAc-transferase genes and determined their chromosomal localization. The coding regions of GALNT1 , -T2 , and -T3 were found to span 11, 16, and 10 exons, respectively. Several intron/exon boundaries were conserved within the three genes. One conserved boundary was shared in a homologous C. elegans GalNAc-transferase gene. Fluorescence in situ hybridization showed that GALNT1 , -T2 , and -T3 are localized at chromosomes 18q12-q21, 1q41-q42, and 2q24-q31, respectively. These results suggest that the members of the polypeptide GalNAc-transferase family diverged early in evolution from a common ancestral gene through gene duplication.