跳转至内容
Merck
  • A New Synthetic Method for the Preparation of Protonated-NHCs and Related Compounds.
A New Synthetic Method for the Preparation of Protonated-NHCs and Related Compounds.
Journal of organometallic chemistry (2006-01-01)
Rodolphe Jazzar, Hongze Liang, Bruno Donnadieu, Guy Bertrand
摘要
Protonated versions of N-heterocyclic carbenes (NHC,H(+)) are classically prepared by closing the ring through the introduction of the CH+ fragment. He we report a totally different synthetic approach, which can be viewed as the addition of a 1,3-diazaallyl anion to a compound featuring two leaving groups (hereafter named "di-electrophile"). Using 1,3- and 1,4-dibromides, six and seven membered NHC,H(+)s have been prepared in good yields. Similarly, with 1,3,2-dioxathiolane-2,2-dioxide as a di-electrophile, imidazolidinium salts were obtained. To illustrate its broad scope of application, this synthetic route has been expanded to the preparation of protonated cyclic amino alkyl carbenes (CAACs) and amino thio carbenes, using 1-aza-allyl and 1,3-azathio-allyl anions, respectively.