跳转至内容
Merck
  • Metabolic fate of the thrombolytic agent benzarone in man: comparison with the rat and dog.
Metabolic fate of the thrombolytic agent benzarone in man: comparison with the rat and dog.
Xenobiotica; the fate of foreign compounds in biological systems (1987-07-01)
S G Wood, B A John, L F Chasseaud, R Bonn, H Grote, K Sandrock, A Darragh, R F Lambe
摘要
1. The metabolic fate of 14C-benzarone in the rat and dog has been compared to that in human subjects. An oral dose was well-absorbed in all three species. However, the 14C excretion patterns differed: humans (100 mg) excreted means of 73 and 19% dose in the urine and faeces respectively, whereas the rat (2 mg/kg) and dog (0.5 mg/kg) excreted greater than 80% in the faeces, mostly during the first 48 h. 2. Much of the faecal 14C was attributable to 14C excreted in the bile which amounted to 59% in the 7 h bile collected from an intravenously dosed dog, and a mean of 72% in the 24 h bile of orally dosed rats. Enterohepatic circulation of 14C was demonstrated in rats. 3. Total 14C in human plasma reached peak concentrations between 1-2 h and declined relatively rapidly, to about 10% of this value within 24 h. Unchanged benzarone was not detected in plasma (less than 25 ng/ml), even after a 400 mg dose, but conjugated benzarone was--accounting for about 10% of the peak concentration of 14C. In the dog, by contrast, conjugated benzarone accounted for about 50% of the peak concentration of 14C of 0.96 microgram equiv./ml at 1 h. The extent of binding of benzarone to human plasma proteins (greater than 99%; in vitro was slightly greater than that (greater than 96%) of total 14C (ex vivo, representing metabolites). 4. Examination of metabolite profiles by h.p.l.c. suggested that in the rat and dog, at least 70% absorbed dose was eliminated by direct conjugation, whereas in humans at least 70% was hydroxylated before conjugation, mainly with glucuronic acid. Hydroxylation occurred in the benzofuran ring and/or the ethyl side-chain. The principal urinary metabolite in humans was the conjugate(s) of the 1-hydroxylated ethyl side-chain derivative (mean 26% dose).
材料
货号
品牌
产品描述
苯扎隆, European Pharmacopoeia (EP) Reference Standard