跳转至内容
Merck
  • Concise synthesis of α-galactosyl ceramide from D-galactosyl iodide and D-lyxose.
Concise synthesis of α-galactosyl ceramide from D-galactosyl iodide and D-lyxose.
Carbohydrate research (2013-01-22)
Yu-Fen Yen, Suvarn S Kulkarni, Chun-Wei Chang, Shun-Yuan Luo
摘要
α-Galactosyl ceramide is synthesized from d-lyxose in 32% overall yield in five steps. The short and efficient protocol involves a one-pot protection and glycosidation of D-lyxose with D-galactosyl iodide as a key step. The α-linked disaccharide obtained was subsequently transformed into α-galactosyl ceramide in four steps involving Z-selective Wittig olefination at C-1, stereo-inversion at C-4 using azide, one-pot reduction of azide and amidation, and global deprotection.
材料
货号
品牌
产品描述
Sigma-Aldrich
D-(-)-来苏糖, 99%
Sigma-Aldrich
(2S,3S,4R)-1-O-(α-D-Galactosyl)-N-hexacosanoyl-2-amino-1,3,4-octadecanetriol, ≥95% (TLC)