跳转至内容
Merck
  • The post-illumination chlorophyll fluorescence transient indicates the RuBP regeneration limitation of photosynthesis in low light in Arabidopsis.
The post-illumination chlorophyll fluorescence transient indicates the RuBP regeneration limitation of photosynthesis in low light in Arabidopsis.
FEBS letters (2010-06-22)
Eiji Gotoh, Yoshichika Kobayashi, Michito Tsuyama
摘要
The mechanism of post-illumination chlorophyll fluorescence transient (PIFT) was investigated in Arabidopsis. PIFT was detected in the wild type after illumination with low light. In the fba3-2 (fructose-1,6-bisphosphate aldolase) mutant, in which PIFT is enhanced, strong light also induced PIFT. PIFT was suppressed not only in the triose phosphate/phosphate translocator (tpt-2) mutant, but also in tpt-2 fba3-2, suggesting that triose phosphates, such as dihydroxyacetone phosphate (DHAP), are involved in the PIFT mechanism. We concluded that PIFT is associated with ribulose-1,5-bisphosphate (RuBP)-regeneration limitation of photosynthesis in low light.
材料
货号
品牌
产品描述
Sigma-Aldrich
L-Ribulose, ≥90% (HPLC)
Sigma-Aldrich
D-核酮糖 溶液, ~1 M in H2O, ≥97.0% (HPLC)