跳转至内容
Merck
  • Differences in the glycosylation of recombinant proteins expressed in HEK and CHO cells.
Differences in the glycosylation of recombinant proteins expressed in HEK and CHO cells.
Journal of biotechnology (2012-07-21)
Amelie Croset, Laurence Delafosse, Jean-Philippe Gaudry, Christian Arod, Loic Glez, Christophe Losberger, Damien Begue, Ana Krstanovic, Flavie Robert, Francis Vilbois, Laurent Chevalet, Bruno Antonsson
摘要
Glycosylation is one of the most common posttranslational modifications of proteins. It has important roles for protein structure, stability and functions. In vivo the glycostructures influence pharmacokinetics and immunogenecity. It is well known that significant differences in glycosylation and glycostructures exist between recombinant proteins expressed in mammalian, yeast and insect cells. However, differences in protein glycosylation between different mammalian cell lines are much less well known. In order to examine differences in glycosylation in mammalian cells we have expressed 12 proteins in the two commonly used cell lines HEK and CHO. The cells were transiently transfected, and the expressed proteins were purified. To identify differences in glycosylation the proteins were analyzed on SDS-PAGE, isoelectric focusing (IEF), mass spectrometry and released glycans on capillary gel electrophoresis (CGE-LIF). For all proteins significant differences in the glycosylation were detected. The proteins migrated differently on SDS-PAGE, had different isoform patterns on IEF, showed different mass peak distributions on mass spectrometry and showed differences in the glycostructures detected in CGE. In order to verify that differences detected were attributed to glycosylation the proteins were treated with deglycosylating enzymes. Although, culture conditions induced minor changes in the glycosylation the major differences were between the two cell lines.
材料
货号
品牌
产品描述
Sigma-Aldrich
糖苷酶F 来源于脑膜脓毒性伊丽莎白菌, BioReagent, ≥95% (SDS-PAGE)
Sigma-Aldrich
糖苷酶F from Elizabethkingia miricola, buffered aqueous solution
Sigma-Aldrich
PNGase F脑膜炎沙门氏菌, ready-to-use solution, recombinant, expressed in E. coli
Sigma-Aldrich
糖苷酶F 来源于脑膜脓毒性伊丽莎白菌, lyophilized powder, recombinant, expressed in E. coli
Sigma-Aldrich
糖苷酶F 来源于脑膜脓毒性伊丽莎白菌, recombinant, expressed in E. coli, set of 100 units nanomolar unit
Sigma-Aldrich
糖肽酶 A 来源于杏仁, buffered aqueous glycerol solution, ≥0.05 unit/mL