跳转至内容
Merck
  • Hypoxia enhances migration and invasion in glioblastoma by promoting a mesenchymal shift mediated by the HIF1α-ZEB1 axis.
Hypoxia enhances migration and invasion in glioblastoma by promoting a mesenchymal shift mediated by the HIF1α-ZEB1 axis.
Cancer letters (2015-01-17)
Justin V Joseph, Siobhan Conroy, Kirill Pavlov, Pallavi Sontakke, Tushar Tomar, Ellie Eggens-Meijer, Veerakumar Balasubramaniyan, Michiel Wagemakers, Wilfred F A den Dunnen, Frank A E Kruyt
摘要
Glioblastoma (GBM) is the most common brain tumor in adults and the mesenchymal GBM subtype was reported to be the most malignant, presenting severe hypoxia and necrosis. Here, we investigated the possible role of a hypoxic microenvironment for inducing a mesenchymal and invasive phenotype. The exposure of non-mesenchymal SNB75 and U87 cells to hypoxia induced a strong change in cell morphology that was accompanied by enhanced invasive capacity and the acquisition of mesenchymal marker expression. Further analyses showed the induction of HIF1α and HIF2α by hypoxia and exposure to digoxin, a cardiac glycoside known to inhibit HIF1/2 expression, was able to prevent hypoxia-induced mesenchymal transition. ShRNA-mediated knockdown of HIF1α, and not HIF2α, prevented this transition, as well as the knockdown of the EMT transcription factor ZEB1. We provide further evidence for a hypoxia-induced mesenchymal shift in GBM primary material by showing co-localization of GLUT1, ZEB1 and the mesenchymal marker YKL40 in hypoxic regions of the tumor. Collectively, our results identify a HIF1α-ZEB1 signaling axis that promotes hypoxia induced mesenchymal shift and invasion in GBM in a cell line dependent fashion.
材料
货号
品牌
产品描述
Sigma-Aldrich
山羊抗兔 IgG 抗体, 2 mg/mL, Chemicon®