跳转至内容
Merck
  • Nerve sprouting and neurogenic inflammation characterize the neurogenic detrusor overactive bladder of patients no longer responsive to drug therapies.
Nerve sprouting and neurogenic inflammation characterize the neurogenic detrusor overactive bladder of patients no longer responsive to drug therapies.
Journal of cellular and molecular medicine (2019-04-05)
Chiara Traini, Giulio Del Popolo, Maria-Simonetta Faussone-Pellegrini, Daniele Guasti, Stefano Catarinicchia, Maria Giuliana Vannucchi
摘要
Urothelium and Lamina Propria (LP) are considered an integrate sensory system which is able to control the detrusor activity. Complete supra-sacral spinal cord lesions cause Neurogenic Detrusor Overactivity (NDO) whose main symptoms are urgency and incontinence. NDO therapy at first consists in anti-muscarinic drugs; secondly, in intra-vesical injection of botulinum toxin. However, with time, all the patients become insensitive to the drugs and decide for cystoplastic surgery. With the aim to get deeper in both NDO and drug's efficacy lack pathogenesis, we investigated the innervation, muscular and connective changes in NDO bladders after surgery by using morphological and quantitative methodologies. Bladder innervation showed a significant global loss associated with an increase in the nerve endings located in the upper LP where a neurogenic inflammation was also present. Smooth muscle cells (SMC) anomalies and fibrosis were found in the detrusor. The increased innervation in the ULP is suggestive for a sprouting and could condition NDO evolution and drug efficacy length. Denervation might cause the SMC anomalies responsible for the detrusor altered contractile activity and intra-cellular traffic and favour the appearance of fibrosis. Inflammation might accelerate these damages. From the clinical point of view, an early anti-inflammatory treatment could positively influence the disease fate.
材料
货号
品牌
产品描述
Sigma-Aldrich
单克隆抗-肌动蛋白,α-平滑肌, clone 1A4, ascites fluid
Sigma-Aldrich
抗蛋白基因产物9.5抗体, from rabbit, purified by affinity chromatography
Sigma-Aldrich
一氧化氮合酶I抗体, serum, Chemicon®