跳转至内容
Merck
  • Src homology region 2 domain-containing phosphatase 1 positively regulates B cell receptor-induced apoptosis by modulating association of B cell linker protein with Nck and activation of c-Jun NH2-terminal kinase.
Src homology region 2 domain-containing phosphatase 1 positively regulates B cell receptor-induced apoptosis by modulating association of B cell linker protein with Nck and activation of c-Jun NH2-terminal kinase.
Journal of immunology (Baltimore, Md. : 1950) (2002-07-05)
Kazuya Mizuno, Yuko Tagawa, Katsuyuki Mitomo, Noriyuki Watanabe, Tatsuo Katagiri, Mami Ogimoto, Hidetaka Yakura
摘要
Src homology region 2 domain-containing phosphatase 1 (SHP-1) is a key mediator in lymphocyte differentiation, proliferation, and activation. We previously showed that B cell linker protein (BLNK) is a physiological substrate of SHP-1 and that B cell receptor (BCR)-induced activation of c-Jun NH(2)-terminal kinase (JNK) is significantly enhanced in cells expressing a form of SHP-1 lacking phosphatase activity (SHP-1-C/S). In this study, we confirmed that SHP-1 also exerts negative regulatory effects on JNK activation in splenic B cells. To further clarify the role of SHP-1 in B cells, we examined how dephosphorylation of BLNK by SHP-1 affects downstream signaling events. When a BLNK mutant (BLNK Delta N) lacking the NH(2)-terminal region, which contains four tyrosine residues, was introduced in SHP-1-C/S-expressing WEHI-231 cells, the enhanced JNK activation was inhibited. Among candidate proteins likely to regulate JNK activation through BLNK, Nck adaptor protein was found to associate with tyrosine-phosphorylated BLNK and this association was more pronounced in SHP-1-C/S-expressing cells. Furthermore, expression of dominant-negative forms of Nck inhibited BCR-induced JNK activation. Finally, BCR-induced apoptosis was suppressed in SHP-1-C/S-expressing cells and coexpression of Nck SH2 mutants or a dominant-negative form of SEK1 reversed this phenotype. Collectively, these results suggest that SHP-1 acts on BLNK, modulating its association with Nck, which in turn negatively regulates JNK activation but exerts a positive effect on apoptosis.