跳转至内容
Merck
Inhibition of bacterial growth by beta-chloro-D-alanine.
Proceedings of the National Academy of Sciences of the United States of America (1974-02-01)
J M Manning, N E Merrifield, W M Jones, E C Gotschlich
摘要
The D- and L-isomers of beta-chloroalanine inhibit the growth of Diplococcus pneumoniae, Streptococcus pyogenes, Bacillus subtilis, and Escherichia coli. With pneumococcus the inhibition by beta-chloro-D-alanine is completely prevented by either D-alanine or D-alanyl-D-alanine, while L-alanine is not effective in preventing the inhibition. The inhibition of growth by beta-chloro-L-alanine is not affected by D-alanine and is only partially prevented by high concentrations of L-alanine. The intracellular free alanine in untreated E. coli and B. subtilis is about 95% in the D-configuration while the free intracellular alanine in both organisms after treatment with beta-chloro-D-alanine is predominantly the L-isomer. These results suggested that the beta-chloroamino acid inactivates alanine racemase (EC 5.1.1.1). Indeed, when extracts of E. coli or B. subtilis were treated with beta-chloro-D-alanine, the activities of alanine racemase and of D-glutamate-D-alanine transaminase were found to be 90-95% inhibited. Studies with mice have shown that beta-chloro-D-alanine is an effective antibacterial agent in vivo againt D. pneumoniae, S. pyogenes, and E. coli.
材料
货号
品牌
产品描述
Sigma-Aldrich
β-Chloro-D-alanine hydrochloride