跳转至内容
Merck
  • RNA activating-double stranded RNA targeting flt-1 promoter inhibits endothelial cell proliferation through soluble FLT-1 upregulation.
RNA activating-double stranded RNA targeting flt-1 promoter inhibits endothelial cell proliferation through soluble FLT-1 upregulation.
PloS one (2018-03-07)
Susie Choi, Hironori Uehara, Yuanyuan Wu, Subrata Das, Xiaohui Zhang, Bonnie Archer, Lara Carroll, Balamurali Krishna Ambati
摘要
Short-activating RNA (saRNA), which targets gene promoters, has been shown to increase the target gene expression. In this study, we describe the use of an saRNA (Flt a-1) to target the flt-1 promoter, leading to upregulation of the soluble isoform of Flt-1 and inhibition of angiogenesis. We demonstrate that Flt a-1 increased sFlt-1 mRNA and protein levels, while reducing VEGF expression. This was associated with suppression of human umbilical vascular endothelial cell (HUVEC) proliferation and cell cycle arrest at the G0/G1 phase. HUVEC migration and tube formation were also suppressed by Flt a-1. An siRNA targeting Flt-1 blocked the effects of Flt a-1. Flt a-1 effects were not mediated via argonaute proteins. However, trichostatin A and 5'-deoxy-5'-(methylthio) adenosine inhibited Flt a-1 effects, indicating that histone acetylation and methylation are mechanistically involved in RNA activation of Flt-1. In conclusion, RNA activation of sFlt-1 can be used to inhibit angiogenesis.
材料
货号
品牌
产品描述
Sigma-Aldrich
抗Ago2抗体,克隆9E8.2, ascites fluid, clone 9E8.2, Upstate®
Sigma-Aldrich
Monoclonal Anti-AGO1 antibody produced in rat, ~1 mg/mL, clone 4B8, purified immunoglobulin