跳转至内容
Merck
  • Bacterial secretion system skews the fate of Legionella-containing vacuoles towards LC3-associated phagocytosis.
Bacterial secretion system skews the fate of Legionella-containing vacuoles towards LC3-associated phagocytosis.
Scientific reports (2017-03-21)
Andree Hubber, Tomoko Kubori, Cevayir Coban, Takeshi Matsuzawa, Michinaga Ogawa, Tsuyoshi Kawabata, Tamotsu Yoshimori, Hiroki Nagai
摘要
The evolutionarily conserved processes of endosome-lysosome maturation and macroautophagy are established mechanisms that limit survival of intracellular bacteria. Similarly, another emerging mechanism is LC3-associated phagocytosis (LAP). Here we report that an intracellular vacuolar pathogen, Legionella dumoffii, is specifically targeted by LAP over classical endocytic maturation and macroautophagy pathways. Upon infection, the majority of L. dumoffii resides in ER-like vacuoles and replicate within this niche, which involves inhibition of classical endosomal maturation. The establishment of the replicative niche requires the bacterial Dot/Icm type IV secretion system (T4SS). Intriguingly, the remaining subset of L. dumoffii transiently acquires LC3 to L. dumoffii-containing vacuoles in a Dot/Icm T4SS-dependent manner. The LC3-decorated vacuoles are bound by an apparently undamaged single membrane, and fail to associate with the molecules implicated in selective autophagy, such as ubiquitin or adaptors. The process requires toll-like receptor 2, Rubicon, diacylglycerol signaling and downstream NADPH oxidases, whereas ULK1 kinase is dispensable. Together, we have discovered an intracellular pathogen, the survival of which in infected cells is limited predominantly by LAP. The results suggest that L. dumoffii is a valuable model organism for examining the mechanistic details of LAP, particularly induced by bacterial infection.
材料
货号
品牌
产品描述
Sigma-Aldrich
抗 早期内涵体抗原 1(N-末端) 兔抗, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution