CESEDKI4ETET
1 of 10
Sigma-Aldrich
WH0010261M1
Monoclonal Anti-IGSF6 antibody produced in mouse
Millipore
MCGBL4S03
Millidisk®屏障过滤器0.22 µm亲水性/疏水性
CN0371E06
Polygard® CN筒式滤芯
KPHLA05FF1
Opticap XL5 Durapore 0.45 µm,无预滤器 3/4 in.TC/TC
MCGL40S03
Millidisk®小柱,带Durapore® 0.22 µm滤膜
CGW372S03
Polysep™II 1.0/0.2 µm 标称,小柱
KVGLA05FF1
Opticap XL5 Durapore 0.22 µm 3/4 in.TC/TC
Z742623
Tube-O-DIALYZER™ Floats
CW0302S03
Milligard® 0.2 µm标称,小柱
CESEDKT4ETET
MILLIDISK 40 HSG T-L TC 1