主要文件
Z741014
Sigma-Aldrich
Aldrich® Essentials round media storage bottle
wide mouth, capacity 1000 mL, GL 45 polypropylene cap with pour ring
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121800
NACRES:
NB.17
物料
borosilicate glass bottle
polypropylene cap
round bottle
接合处
Joints threaded neck (GL45)
特点
graduations
with cap
包装
pkg of 10 ea
制造商/商品名称
Sigma-Aldrich
容量
1000 mL
正在寻找类似产品? 访问 产品对比指南
一般描述
Borosilicate glass, wide mouth, media/reagent bottles with polypropylene GL 45 thread screw cap with pour ring. Large marking spot and reference graduations.
法律信息
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
历史批次信息供参考:
分析证书(COA)