Key Documents
Z568716
Sigma-Aldrich
Aldrich® combustion apparatus black paper sample wrappers
sample L × W 32 mm × 30 mm, pkg of 100 ea
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121800
NACRES:
NB.43
包裝
pkg of 100 ea
一般說明
Absorb heat quickly for infrared ignition. Sample area 32 x 30 mm, with 8 x 38 mm extending strip as fuse for ignition.
法律資訊
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC