Key Documents
Z553093
Sigma-Aldrich
Aldrich® Buchner funnel with joint and SafetyBarb® hose connector
capacity 60 mL, porosity 10-20 μm
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121809
NACRES:
NB.43
接合處
joint ST/NS24/40
容量
60 mL
孔徑
10-20 μm porosity
正在寻找类似产品? 访问 产品对比指南
一般說明
Order replacement SafetyBarb hose connectors Z547786 (straight) or Z547883 (angled).
法律資訊
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany