Key Documents
Z268208
Waste Solvent Container
Nalgene rectangular D.O.T. carboy, capacity 20 L,
closure size 70 mm
别名:
bottle, carboy
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121800
NACRES:
NB.43
包裝
pack of 1 ea
製造商/商標名
Nalgene 2214-0050
容量
20 L
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
Various sizes of Nalgene bottles or carboys to fit waste solvent funnels. Caps not included.