Key Documents
BR23811
BRAND®紧凑型自动SILBERBRAND滴定管,无支架/夹子
volume 50 mL, with LDPE bottle and PVC filling tube
别名:
SILBERBRAND, 滴定管, 紧凑型自动滴定管, 自动滴定管
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121808
NACRES:
NB.43
材料
AR-Glass
製造商/商標名
BRAND 23811
體積測量範圍
50 mL, accuracy: ±0.10 mL
容積
50 mL
正在寻找类似产品? 访问 产品对比指南
一般說明
The Modular Automatic Burette Concept! Fast to dismantle and easy to clean. Easy to repair - all individual components replaceable! Boro 3.3, detachable PTFE stopcock. Error limits according to class B, DIN EN ISO 385. Calibrated to deliver (TD, Ex).
法律資訊
BRAND is a registered trademark of BRAND GMBH + CO KG