Merck
  • CCR5 antagonists as anti-HIV-1 agents. 1. Synthesis and biological evaluation of 5-oxopyrrolidine-3-carboxamide derivatives.
CCR5 antagonists as anti-HIV-1 agents. 1. Synthesis and biological evaluation of 5-oxopyrrolidine-3-carboxamide derivatives.
Chemical & pharmaceutical bulletin (2004-01-08)
Shinichi Imamura, Yuji Ishihara, Taeko Hattori, Osamu Kurasawa, Yoshihiro Matsushita, Yoshihiro Sugihara, Naoyuki Kanzaki, Yuji Iizawa, Masanori Baba, Shohei Hashiguchi
ABSTRACT
A novel lead compound, N-(3-[4-(4-fluorobenzoyl)piperidin-1-yl]propyl)-1-methyl-5-oxo-N-phenylpyrrolidine-3-carboxamide (1), was identified as a CCR5 antagonist by high-throughput screening using [(125)I]RANTES and CCR5-expressing CHO cells. The IC(50) value of 1 was 1.9 microM. In an effort to improve the binding affinity of 1, a series of 5-oxopyrrolidine-3-carboxamides was synthesized. Introduction of 3,4-dichloro substituents to the central phenyl ring (10i, IC(50)=0.057 microM; 11b, IC(50)=0.050 microM) or replacing the 1-methyl group of the 5-oxopyrrolidine moiety with a 1-benzyl group (12e, IC(50)=0.038 microM) was found to be effective for improving CCR5 affinity. Compound 10i, 11b, and 12e also inhibited CCR5-using HIV-1 envelope-mediated membrane fusion with IC(50) values of 0.44, 0.19, and 0.49 microM, respectively.
Sigma-Aldrich
2,4-Dimethoxybenzylamine, 98%