Merck
  • Methylation of Gata3 protein at Arg-261 regulates transactivation of the Il5 gene in T helper 2 cells.
Methylation of Gata3 protein at Arg-261 regulates transactivation of the Il5 gene in T helper 2 cells.
The Journal of biological chemistry (2015-04-12)
Hiroyuki Hosokawa, Miki Kato, Hiroyuki Tohyama, Yuuki Tamaki, Yusuke Endo, Motoko Y Kimura, Damon John Tumes, Shinichiro Motohashi, Masaki Matsumoto, Keiichi I Nakayama, Tomoaki Tanaka, Toshinori Nakayama
ABSTRACT
Gata3 acts as a master regulator for T helper 2 (Th2) cell differentiation by inducing chromatin remodeling of the Th2 cytokine loci, accelerating Th2 cell proliferation, and repressing Th1 cell differentiation. Gata3 also directly transactivates the interleukin-5 (Il5) gene via additional mechanisms that have not been fully elucidated. We herein identified a mechanism whereby the methylation of Gata3 at Arg-261 regulates the transcriptional activation of the Il5 gene in Th2 cells. Although the methylation-mimicking Gata3 mutant retained the ability to induce IL-4 and repress IFNγ production, the IL-5 production was selectively impaired. We also demonstrated that heat shock protein (Hsp) 60 strongly associates with the methylation-mimicking Gata3 mutant and negatively regulates elongation of the Il5 transcript by RNA polymerase II. Thus, arginine methylation appears to play a pivotal role in the organization of Gata3 complexes and the target gene specificity of Gata3.
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
MISSION® esiRNA, targeting human HSPD1