- Design, synthesis and evaluation of diazadibenzocrown ethers as Pb(2+) extractants and carriers in plasticized cellulose triacetate membranes.
Design, synthesis and evaluation of diazadibenzocrown ethers as Pb(2+) extractants and carriers in plasticized cellulose triacetate membranes.
Talanta (2008-10-31)
J C Aguilar, E R Miguel, J Gyves, R A Bartsch, M Kim
PMID18968340
ABSTRACT
The ligands 4,7-diaza-2,3,8,9-dibenzo-15-crown-5 (L1), 4,10-diaza-2,3,11,12-dibenzo-18-crown-6 (L2), 4,10-diaza-2,3,11,12-di(4'-tert-butylbenzo)-18-crown-6 (L3) and N,N-di(methylenecarboxyethoxy) 4,10-diaza-2,3,11,12-dibenzo-18-crown-6 (L4) have been prepared. Partition coefficients and acid dissociation constants for these four diazadibenzocrown ether compounds were determined in water-chloroform. Their effectiveness was assessed in solvent extraction of Pb(2+) from aqueous solutions into toluene. Ligands L3 and L4 provide high selectivity for Pb(2+) over Cd(2+) and Zn(2+) in transport across plasticized cellulose triacetate membranes.
MATERIALS