Merck
  • Identification of myopia-associated WNT7B polymorphisms provides insights into the mechanism underlying the development of myopia.
Identification of myopia-associated WNT7B polymorphisms provides insights into the mechanism underlying the development of myopia.
Nature communications (2015-04-01)
Masahiro Miyake, Kenji Yamashiro, Yasuharu Tabara, Kenji Suda, Satoshi Morooka, Hideo Nakanishi, Chiea-Chuen Khor, Peng Chen, Fan Qiao, Isao Nakata, Yumiko Akagi-Kurashige, Norimoto Gotoh, Akitaka Tsujikawa, Akira Meguro, Sentaro Kusuhara, Ozen Polasek, Caroline Hayward, Alan F Wright, Harry Campbell, Andrea J Richardson, Maria Schache, Masaki Takeuchi, David A Mackey, Alex W Hewitt, Gabriel Cuellar, Yi Shi, Luling Huang, Zhenglin Yang, Kim Hung Leung, Patrick Y P Kao, Maurice K H Yap, Shea Ping Yip, Muka Moriyama, Kyoko Ohno-Matsui, Nobuhisa Mizuki, Stuart MacGregor, Veronique Vitart, Tin Aung, Seang-Mei Saw, E-Shyong Tai, Tien Yin Wong, Ching-Yu Cheng, Paul N Baird, Ryo Yamada, Fumihiko Matsuda, Nagahisa Yoshimura
ABSTRACT
Myopia can cause severe visual impairment. Here, we report a two-stage genome-wide association study for three myopia-related traits in 9,804 Japanese individuals, which was extended with trans-ethnic replication in 2,674 Chinese and 2,690 Caucasian individuals. We identify WNT7B as a novel susceptibility gene for axial length (rs10453441, Pmeta=3.9 × 10(-13)) and corneal curvature (Pmeta=2.9 × 10(-40)) and confirm the previously reported association between GJD2 and myopia. WNT7B significantly associates with extreme myopia in a case-control study with 1,478 Asian patients and 4,689 controls (odds ratio (OR)meta=1.13, Pmeta=0.011). We also find in a mouse model of myopia downregulation of WNT7B expression in the cornea and upregulation in the retina, suggesting its possible role in the development of myopia.
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Anti-Brn-3a Antibody, POU-domain protein, clone 5A3.2, culture supernatant, clone 5A3.2, Chemicon®