Merck
  • Tight confinement of semiconductor quantum dots within zeolite by surface silylation.
Tight confinement of semiconductor quantum dots within zeolite by surface silylation.
Langmuir : the ACS journal of surfaces and colloids (2005-06-15)
Nak Cheon Jeong, Hyun Sung Kim, Kyung Byung Yoon
ABSTRACT
Various MX (M = Cd, Zn, and Mn, X = S and Se) semiconductor quantum dots (QDs) were prepared in zeolite Y. While the QDs are readily expelled from zeolite interior upon exposure of the MX QD-incorporating zeolite Y ([MX]-Y) to the ambient atmosphere due to moisture adsorption, they remain tightly confined within zeolites even after exposure to the moist atmosphere for several weeks when the surfaces were silylated with various silylating agents. This methodology will facilitate the characterization of the zeolite-encapsulated QDs and the application of QD-incorporating zeolites.
Sigma-Aldrich
Aluminum sulfate octadecahydrate, puriss., meets analytical specification of Ph. Eur., BP, 100-110%, 51.0-59.0% Al2(SO4)3 basis