- Methods and reagents. Agarose gel electrophoresis in your kitchen.
Methods and reagents. Agarose gel electrophoresis in your kitchen.
Trends in biochemical sciences (1995-05-01)
P N Hengen
PMID7610485
ABSTRACT
Methods and reagents is a unique monthly column that highlights current discussions in the newsgroup bionet.molbio.methods-reagents, available on the Internet. This month's column describes how to make an electrophoresis system from common household items. For details on how to partake in the newsgroup, see the accompanying box.
MATERIALS